Product Details

SNP ID
rs41292543
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:46309111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTCCTTTTGTAGGAGGAAGAGG[A/G]AGAGGAGGAATTAGTGGTAAGAACT
Phenotype
MIM: 613844
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC41 PubMed Links

Gene Details

Gene
LRRC41
Gene Name
leucine rich repeat containing 41
There are no transcripts associated with this gene.

Gene
UQCRH
Gene Name
ubiquinol-cytochrome c reductase hinge protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297565.1 219 UTR 5 NP_001284494.1
NM_001297566.1 219 Intron NP_001284495.1
NM_006004.3 219 Missense Mutation GAA,GGA E22G NP_005995.2

View Full Product Details