Product Details

SNP ID
rs61774748
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:41375831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGGCTGGAGGCATAAGATTAGA[G/T]GAGGTTGAGCTGTTGGAGAATGAAG
Phenotype
MIM: 611457
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FOXO6 PubMed Links

Gene Details

Gene
FOXO6
Gene Name
forkhead box O6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291281.2 Intron NP_001278210.2
Gene
LOC101929901
Gene Name
uncharacterized LOC101929901
There are no transcripts associated with this gene.

View Full Product Details