Product Details

SNP ID
rs73077325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2041924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCTAGGTTCCTCTAAACCCGCG[A/G]CCAGCGCAGTCTCGGGTTTCCATGA
Phenotype
MIM: 614690
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C4orf48 PubMed Links

Gene Details

Gene
C4orf48
Gene Name
chromosome 4 open reading frame 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001141936.2 Intron NP_001135408.2
NM_001168243.1 Intron NP_001161715.1
XM_011513469.2 Intron XP_011511771.1
XM_011513471.2 Intron XP_011511773.1
XM_017008190.1 Intron XP_016863679.1

View Full Product Details