Product Details

SNP ID
rs78986900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179912839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACTTGCACTTAATTGGAGATAAA[A/G]GCTGTTCCAAAAGCAAATTTGAATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF130 PubMed Links

Gene Details

Gene
RNF130
Gene Name
ring finger protein 130
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280801.1 9166 Intron NP_001267730.1
NM_018434.5 9166 Intron NP_060904.2
XM_011534593.2 9166 UTR 3 XP_011532895.1
Gene
TBC1D9B
Gene Name
TBC1 domain family member 9B
There are no transcripts associated with this gene.

View Full Product Details