Product Details

SNP ID
rs112012998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:8791374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAATAATTGTCTTTAGTGTCCTT[A/G]GCAGCCTAAAAGAAAAAGTCTAGTT
Phenotype
MIM: 300477
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM9A PubMed Links

Gene Details

Gene
FAM9A
Gene Name
family with sequence similarity 9 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171186.1 Intron NP_001164657.1
NM_174951.3 Intron NP_777611.1

View Full Product Details