Product Details

SNP ID
rs377564688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10270985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGGAGGATGACCCTCTCGGCCC[C/G]GGCACCCTGTCAGTCCGGAAATAAC
Phenotype
MIM: 147840
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ICAM1 PubMed Links

Gene Details

Gene
ICAM1
Gene Name
intercellular adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000201.2 145 UTR 5 NP_000192.2

View Full Product Details