Product Details

SNP ID
rs146529730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81919357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAGCCGCTCAGGGCCGTGGGCC[A/G]GGCCCTGATCACCAGTCAGAAGTGT
Phenotype
MIM: 602020 MIM: 602679 MIM: 606212
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAFG PubMed Links

Gene Details

Gene
MAFG
Gene Name
MAF bZIP transcription factor G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002359.3 3950 UTR 3 NP_002350.1
NM_032711.3 3950 UTR 3 NP_116100.2
XM_011523578.1 3950 UTR 3 XP_011521880.1
Gene
MAFG-AS1
Gene Name
MAFG antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
PCYT2
Gene Name
phosphate cytidylyltransferase 2, ethanolamine
There are no transcripts associated with this gene.

Gene
SIRT7
Gene Name
sirtuin 7
There are no transcripts associated with this gene.

View Full Product Details