Product Details
- SNP ID
-
hCV27831997
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:61453740 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATAATGACAAAGTAACAAGATAC[C/T]GTCGGGGTGTGTTGAAGAATAATAC
- Phenotype
-
MIM: 601719
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TBX4
PubMed Links
Gene Details
- Gene
- TBX4
- Gene Name
- T-box 4
View Full Product Details