Product Details

SNP ID
rs77405826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:101047166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTATGGCATCTTGCTTCCCTGGT[A/G]CTTAAAGAGAGGTCAAACTTGGTGC
Phenotype
MIM: 610164 MIM: 610959 MIM: 610960 MIM: 610961 MIM: 610983 MIM: 615385 MIM: 615037
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR1185-1 PubMed Links

Gene Details

Gene
MIR1185-1
Gene Name
microRNA 1185-1
There are no transcripts associated with this gene.

Gene
MIR1185-2
Gene Name
microRNA 1185-2
There are no transcripts associated with this gene.

Gene
MIR134
Gene Name
microRNA 134
There are no transcripts associated with this gene.

Gene
MIR300
Gene Name
microRNA 300
There are no transcripts associated with this gene.

Gene
MIR323B
Gene Name
microRNA 323b
There are no transcripts associated with this gene.

Gene
MIR376A1
Gene Name
microRNA 376a-1
There are no transcripts associated with this gene.

Gene
MIR376A2
Gene Name
microRNA 376a-2
There are no transcripts associated with this gene.

Gene
MIR376B
Gene Name
microRNA 376b
There are no transcripts associated with this gene.

Gene
MIR376C
Gene Name
microRNA 376c
There are no transcripts associated with this gene.

Gene
MIR381
Gene Name
microRNA 381
There are no transcripts associated with this gene.

Gene
MIR381HG
Gene Name
MIR381 host gene
There are no transcripts associated with this gene.

Gene
MIR382
Gene Name
microRNA 382
There are no transcripts associated with this gene.

Gene
MIR485
Gene Name
microRNA 485
There are no transcripts associated with this gene.

Gene
MIR487A
Gene Name
microRNA 487a
There are no transcripts associated with this gene.

Gene
MIR487B
Gene Name
microRNA 487b
There are no transcripts associated with this gene.

Gene
MIR539
Gene Name
microRNA 539
There are no transcripts associated with this gene.

Gene
MIR544A
Gene Name
microRNA 544a
There are no transcripts associated with this gene.

Gene
MIR654
Gene Name
microRNA 654
There are no transcripts associated with this gene.

Gene
MIR655
Gene Name
microRNA 655
There are no transcripts associated with this gene.

Gene
MIR668
Gene Name
microRNA 668
There are no transcripts associated with this gene.

Gene
MIR889
Gene Name
microRNA 889
There are no transcripts associated with this gene.

View Full Product Details