Product Details
- SNP ID
-
rs4674470
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:219906502 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGATGTCACACAGTGAAGATGCCCC[C/T]AGGTGACATCACATCCTGAGAGGAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4268
PubMed Links
Gene Details
- Gene
- MIR4268
- Gene Name
- microRNA 4268
There are no transcripts associated with this gene.
View Full Product Details