Product Details

SNP ID
rs191254669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69915129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGCTCTGCTCCGGGTTCTGTC[A/C]CTGTGTCGGCGGTGCCCAGCTCACT
Phenotype
MIM: 600021
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MXD1 PubMed Links

Gene Details

Gene
MXD1
Gene Name
MAX dimerization protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202513.1 89 UTR 5 NP_001189442.1
NM_001202514.1 89 UTR 5 NP_001189443.1
NM_002357.3 89 UTR 5 NP_002348.1
Gene
SNRNP27
Gene Name
small nuclear ribonucleoprotein U4/U6.U5 subunit 27
There are no transcripts associated with this gene.

View Full Product Details