Product Details

SNP ID
rs531365257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:88562953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAGGCGTGCGCAGGCGCGGACA[C/G]GGGCGCATGCGCGTTCACTCCGGCG
Phenotype
MIM: 603271 MIM: 613279
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PTPN21 PubMed Links

Gene Details

Gene
PTPN21
Gene Name
protein tyrosine phosphatase, non-receptor type 21
There are no transcripts associated with this gene.

Gene
ZC3H14
Gene Name
zinc finger CCCH-type containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160103.1 40 UTR 5 NP_001153575.1
NM_001160104.1 40 UTR 5 NP_001153576.1
NM_001326295.1 40 UTR 5 NP_001313224.1
NM_001326296.1 40 UTR 5 NP_001313225.1
NM_001326297.1 40 UTR 5 NP_001313226.1
NM_001326298.1 40 UTR 5 NP_001313227.1
NM_001326299.1 40 UTR 5 NP_001313228.1
NM_001326300.1 40 UTR 5 NP_001313229.1
NM_001326301.1 40 UTR 5 NP_001313230.1
NM_001326302.1 40 UTR 5 NP_001313231.1
NM_001326303.1 40 UTR 5 NP_001313232.1
NM_001326304.1 40 UTR 5 NP_001313233.1
NM_001326305.1 40 UTR 5 NP_001313234.1
NM_001326306.1 40 UTR 5 NP_001313235.1
NM_001326307.1 40 UTR 5 NP_001313236.1
NM_001326308.1 40 UTR 5 NP_001313237.1
NM_001326309.1 40 UTR 5 NP_001313238.1
NM_001326310.1 40 UTR 5 NP_001313239.1
NM_001326311.1 40 UTR 5 NP_001313240.1
NM_001326312.1 40 UTR 5 NP_001313241.1
NM_001326313.1 40 UTR 5 NP_001313242.1
NM_001326314.1 40 UTR 5 NP_001313243.1
NM_001326315.1 40 UTR 5 NP_001313244.1
NM_001326316.1 40 Intron NP_001313245.1
NM_024824.4 40 UTR 5 NP_079100.2
NM_207660.3 40 UTR 5 NP_997543.1
NM_207661.2 40 Intron NP_997544.1
NM_207662.3 40 Intron NP_997545.2
XM_005268067.4 40 UTR 5 XP_005268124.1
XM_005268068.4 40 UTR 5 XP_005268125.1
XM_005268069.4 40 UTR 5 XP_005268126.1
XM_005268070.4 40 UTR 5 XP_005268127.1
XM_005268071.4 40 UTR 5 XP_005268128.1
XM_005268073.3 40 Intron XP_005268130.1
XM_006720257.2 40 Intron XP_006720320.1
XM_011537160.2 40 Intron XP_011535462.1
XM_011537161.2 40 UTR 5 XP_011535463.1
XM_011537162.2 40 UTR 5 XP_011535464.1
XM_017021648.1 40 UTR 5 XP_016877137.1
XM_017021649.1 40 UTR 5 XP_016877138.1
XM_017021650.1 40 UTR 5 XP_016877139.1

View Full Product Details