Product Details

SNP ID
rs113876423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87155643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCATTTTGTGAAGAAAATTCCT[A/G]TAATAGAAATCACAAATAGATTTTG
Phenotype
MIM: 608491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DMTF1 PubMed Links

Gene Details

Gene
DMTF1
Gene Name
cyclin D binding myb like transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142326.1 Intron NP_001135798.1
NM_001142327.1 Intron NP_001135799.1
NM_021145.3 Intron NP_066968.3
XM_011516735.2 Intron XP_011515037.1
XM_011516737.1 Intron XP_011515039.1
XM_011516739.2 Intron XP_011515041.1
XM_017012865.1 Intron XP_016868354.1
XM_017012866.1 Intron XP_016868355.1
XM_017012867.1 Intron XP_016868356.1
XM_017012868.1 Intron XP_016868357.1
XM_017012869.1 Intron XP_016868358.1
XM_017012870.1 Intron XP_016868359.1
XM_017012871.1 Intron XP_016868360.1
XM_017012872.1 Intron XP_016868361.1
XM_017012873.1 Intron XP_016868362.1
XM_017012874.1 Intron XP_016868363.1
XM_017012875.1 Intron XP_016868364.1
XM_017012876.1 Intron XP_016868365.1
XM_017012877.1 Intron XP_016868366.1
XM_017012878.1 Intron XP_016868367.1
Gene
LOC101927420
Gene Name
uncharacterized LOC101927420
There are no transcripts associated with this gene.

View Full Product Details