Product Details
- SNP ID
-
rs373028668
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:74958673 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCGGCCGCGCGCAGCACGCTCAA[A/G]GCCGGGATGGCGGCGGCGGCGGCGG
- Phenotype
-
MIM: 607269
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105374809
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs528770146] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105374809
- Gene Name
- uncharacterized LOC105374809
There are no transcripts associated with this gene.
- Gene
- POLE4
- Gene Name
- DNA polymerase epsilon 4, accessory subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019896.2 |
26 |
UTR 5 |
|
|
NP_063949.2 |
View Full Product Details