Product Details

SNP ID
rs57938858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43520259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAGGGAAGTGAGGGTCATCATC[C/G]CAGGGGCCCTCTCTCCAGCTGAGCC
Phenotype
MIM: 600178 MIM: 605230
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAP1A PubMed Links

Gene Details

Gene
MAP1A
Gene Name
microtubule associated protein 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002373.5 Intron NP_002364.5
XM_017022189.1 Intron XP_016877678.1
Gene
TP53BP1
Gene Name
tumor protein p53 binding protein 1
There are no transcripts associated with this gene.

View Full Product Details