Product Details

SNP ID
rs28533138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:8319254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGGATGTTAAAGTGCACGGGCA[C/T]GGACGGGCTGCAGTAGGAGACTGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SGK223 PubMed Links

Gene Details

Gene
SGK223
Gene Name
homolog of rat pragma of Rnd2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080826.2 3796 Missense Mutation ATG,GTG M1041V NP_001074295.2
XM_005272369.4 3796 Missense Mutation ATG,GTG M1041V XP_005272426.2
XM_005272370.4 3796 Missense Mutation ATG,GTG M1041V XP_005272427.2
XM_011543813.2 3796 Intron XP_011542115.1

View Full Product Details