Product Details

SNP ID
rs8177123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:142860375 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACAGTATTTCTAAGGACTCCGTGA[C/T]TCTGGTTTGCTTCAGTGTGTCAAGC
Phenotype
MIM: 602757
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPHB6 PubMed Links

Gene Details

Gene
EPHB6
Gene Name
EPH receptor B6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280794.2 Intron NP_001267723.2
NM_001280795.2 Intron NP_001267724.2
NM_004445.5 Intron NP_004436.4
XM_006715881.2 Intron XP_006715944.1
XM_011515879.1 Intron XP_011514181.1
XM_011515880.1 Intron XP_011514182.1
XM_011515881.2 Intron XP_011514183.1
XM_011515882.2 Intron XP_011514184.1

View Full Product Details