Product Details

SNP ID
rs41311479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:12975167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCTCGTACTCGTGCGCCTCGCTT[C/T]GGTGAGCCCCAGGGCCCCTGCCTCC
Phenotype
MIM: 300159
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMSB4X PubMed Links

Gene Details

Gene
TMSB4X
Gene Name
thymosin beta 4, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021109.3 Intron NP_066932.1

View Full Product Details