Product Details

SNP ID
hCV27837465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:70172438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGTTATTAGCAATGGAAATAAG[A/C]CAATAATAGTCTCTCAATACAGGCT
Phenotype
MIM: 600681
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCNJ2 PubMed Links

Gene Details

Gene
KCNJ2
Gene Name
potassium voltage-gated channel subfamily J member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000891.2 Intron NP_000882.1
Gene
KCNJ2-AS1
Gene Name
KCNJ2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details