Product Details

SNP ID
rs113457902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:23299165 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAAGCAAGGTGGGCAAGCAACAC[C/T]AGTTCTTTATTCCCGCACGCCCTAA
Phenotype
MIM: 614624
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MALSU1 PubMed Links

Gene Details

Gene
MALSU1
Gene Name
mitochondrial assembly of ribosomal large subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138446.1 Intron NP_612455.1
XM_017011731.1 Intron XP_016867220.1

View Full Product Details