Product Details

SNP ID
rs144264470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:151737127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTTCAGAGGCTGGAACAAGCTT[A/G]GGCGGGGGTTGAGGGAGCTCCAGGG
Phenotype
MIM: 300338
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNGA2 PubMed Links

Gene Details

Gene
CNGA2
Gene Name
cyclic nucleotide gated channel alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005140.1 Intron NP_005131.1

View Full Product Details