Product Details

SNP ID
rs558816396
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:153609085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATCTGGATCTACTCTGTGAGTCA[C/T]GGCCCATGCCCAAAACCCCCCCACA
Phenotype
MIM: 601989 MIM: 607986
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
S100A13 PubMed Links

Gene Details

Gene
S100A13
Gene Name
S100 calcium binding protein A13
There are no transcripts associated with this gene.

Gene
S100A14
Gene Name
S100 calcium binding protein A14
There are no transcripts associated with this gene.

Gene
S100A16
Gene Name
S100 calcium binding protein A16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317007.1 271 UTR 5 NP_001303936.1
NM_001317008.1 271 Intron NP_001303937.1
NM_080388.2 271 Intron NP_525127.1

View Full Product Details