Product Details

SNP ID
rs74090086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104868311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGAGGCTGGAGCCCAGCTTCTC[C/T]GGAAGCTGCCAGCTTCCCTTAGAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP170B PubMed Links

Gene Details

Gene
CEP170B
Gene Name
centrosomal protein 170B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112726.2 Intron NP_001106197.1
NM_015005.2 Intron NP_055820.2
XM_005267550.3 Intron XP_005267607.1
XM_011536665.2 Intron XP_011534967.1
XM_011536666.2 Intron XP_011534968.1
XM_011536667.2 Intron XP_011534969.1
XM_017021227.1 Intron XP_016876716.1
XM_017021228.1 Intron XP_016876717.1
XM_017021229.1 Intron XP_016876718.1
XM_017021230.1 Intron XP_016876719.1

View Full Product Details