Product Details

SNP ID
rs10974727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:4680407 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGACAACTGAGTAAATTCTTTAAA[C/T]CTCGCTTTTGACGCTGGCGTACACC
Phenotype
MIM: 610346
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC37L1 PubMed Links

Gene Details

Gene
CDC37L1
Gene Name
cell division cycle 37 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017913.3 Intron NP_060383.2
Gene
CDC37L1-AS1
Gene Name
CDC37L1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details