Product Details
- SNP ID
-
hCV27841498
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:123671125 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGGTGGTTGCCATAGCTCCGGGC[A/G]TTCGCTTGCAAGATGGCGGCGGCGG
- Phenotype
-
MIM: 601750
MIM: 613846
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GTF2H3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78846567] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GTF2H3
- Gene Name
- general transcription factor IIH subunit 3
There are no transcripts associated with this gene.
- Gene
- TCTN2
- Gene Name
- tectonic family member 2
View Full Product Details