Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_005892.3 | 13 | Intron | NP_005883.2 | ||
XM_006722062.3 | 13 | Intron | XP_006722125.1 | ||
XM_006722063.3 | 13 | Intron | XP_006722126.1 | ||
XM_006722064.3 | 13 | Intron | XP_006722127.1 | ||
XM_006722065.3 | 13 | Intron | XP_006722128.1 | ||
XM_006722066.3 | 13 | Intron | XP_006722129.1 | ||
XM_006722069.2 | 13 | Intron | XP_006722132.1 | ||
XM_006722070.3 | 13 | Intron | XP_006722133.1 | ||
XM_011525179.2 | 13 | Intron | XP_011523481.1 | ||
XM_011525180.2 | 13 | UTR 5 | XP_011523482.1 | ||
XM_011525182.2 | 13 | Intron | XP_011523484.1 |