Product Details

SNP ID
rs574274228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10316759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATAGTCAAGCTCCCAGAGCTTG[C/G]CATCTGTGGCTCTGGCCAGCAGGGC
Phenotype
MIM: 614585 MIM: 609950 MIM: 611639
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FDX1L PubMed Links

Gene Details

Gene
FDX1L
Gene Name
ferredoxin 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031734.3 3046 Intron NP_001026904.2
Gene
RAVER1
Gene Name
ribonucleoprotein, PTB binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133452.2 3046 UTR 3 NP_597709.2
XM_011527671.1 3046 UTR 3 XP_011525973.1
Gene
ZGLP1
Gene Name
zinc finger, GATA-like protein 1
There are no transcripts associated with this gene.

View Full Product Details