Product Details

SNP ID
rs544776973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:57228468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCCGGAGCCCAAGACCGCTCTC[C/G]CCGCCGAATGTGCCCGTGTCGCGGC
Phenotype
MIM: 604299 MIM: 601802
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APPL1 PubMed Links

Gene Details

Gene
APPL1
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012096.2 Intron NP_036228.1
XM_011533583.2 Intron XP_011531885.1
Gene
HESX1
Gene Name
HESX homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003865.2 Intron NP_003856.1
XM_005265526.4 Intron XP_005265583.1
XM_006713379.3 Intron XP_006713442.1
XM_011534204.2 Intron XP_011532506.1
XM_011534205.2 Intron XP_011532507.1
XM_017007421.1 Intron XP_016862910.1

View Full Product Details