Product Details
- SNP ID
-
rs62118143
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:16500716 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTTGCAAAAGACAGTGTGAGCTTT[A/T]GCCAAGTAGGAGCCATTGAATGCTC
- Phenotype
-
MIM: 611414
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C19orf44
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs566396315] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C19orf44
- Gene Name
- chromosome 19 open reading frame 44
- Gene
- CALR3
- Gene Name
- calreticulin 3
There are no transcripts associated with this gene.
View Full Product Details