Product Details

SNP ID
rs79877835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90874152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGGACCTCCCCTAGCTTCCCCC[A/C]CTTCCTCCAGGGCCTTGGCTTCTGG
Phenotype
MIM: 136950
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FURIN PubMed Links

Gene Details

Gene
FURIN
Gene Name
furin, paired basic amino acid cleaving enzyme
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289823.1 Intron NP_001276752.1
NM_001289824.1 Intron NP_001276753.1
NM_002569.3 Intron NP_002560.1

View Full Product Details