Product Details

SNP ID
rs140501445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:45095277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATGGATGAATACAGAAATTCTA[A/G]TAATGGCAGCACAGGCAACAGTTCA
Phenotype
MIM: 614907
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRPF39 PubMed Links

Gene Details

Gene
PRPF39
Gene Name
pre-mRNA processing factor 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017922.3 208 Missense Mutation AAT,AGT N13S NP_060392.3

View Full Product Details