Product Details

SNP ID
rs150882458
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:11771387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAATCCGACATAAAATAAACTGT[C/T]CAGAAGAGGGTAGGGGGACTCAGCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf167 PubMed Links

Gene Details

Gene
C1orf167
Gene Name
chromosome 1 open reading frame 167
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010881.1 Intron NP_001010881.1
XM_011541269.2 Intron XP_011539571.2
XM_011541271.2 Intron XP_011539573.2
XM_011541272.2 Intron XP_011539574.1
XM_011541274.2 Intron XP_011539576.2
XM_011541275.2 Intron XP_011539577.2
XM_011541276.2 Intron XP_011539578.1
XM_011541277.2 Intron XP_011539579.1
XM_011541278.2 Intron XP_011539580.1
XM_011541280.2 Intron XP_011539582.2
XM_011541281.2 Intron XP_011539583.2
XM_017001062.1 Intron XP_016856551.1
XM_017001063.1 Intron XP_016856552.1
XM_017001064.1 Intron XP_016856553.1
Gene
LOC102724659
Gene Name
uncharacterized LOC102724659
There are no transcripts associated with this gene.

View Full Product Details