Product Details

SNP ID
rs528569972
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:52404551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCATGACGTACATCCGCTAACA[C/T]GCAGCCGCCATCATTGATTCGATAG
Phenotype
MIM: 601902 MIM: 617031
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ORC1 PubMed Links

Gene Details

Gene
ORC1
Gene Name
origin recognition complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190818.1 5 Intron NP_001177747.1
NM_001190819.1 5 Intron NP_001177748.1
NM_004153.3 5 Intron NP_004144.2
XM_011541527.2 5 Intron XP_011539829.1
XM_017001388.1 5 Intron XP_016856877.1
XM_017001389.1 5 Intron XP_016856878.1
Gene
PRPF38A
Gene Name
pre-mRNA processing factor 38A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032864.3 5 UTR 5 NP_116253.2
XM_011542315.2 5 UTR 5 XP_011540617.1

View Full Product Details