Product Details

SNP ID
rs10896272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56346117 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTCTCATGGGGATTACAGACAAC[A/C]CTGGGCTGCAGGCTCCACTGTTTGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR8K1 PubMed Links

Gene Details

Gene
OR8K1
Gene Name
olfactory receptor family 8 subfamily K member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002907.1 79 Missense Mutation ACT,CCT T27P NP_001002907.1

View Full Product Details