Product Details
- SNP ID
-
rs114744773
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:96969378 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAATACAAGTAAATGCAAGTAAAT[C/T]TGCATGCATAAAGCATGTAGAATAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DLX6-AS1
PubMed Links
Gene Details
- Gene
- DLX6-AS1
- Gene Name
- DLX6 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details