Product Details

SNP ID
rs77787773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56315868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCCCAAGCGCTGGAAGACCGCT[A/G]GACTCTCACTTTGGCCCCAGTGGCT
Phenotype
MIM: 605861
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNPY2 PubMed Links

Gene Details

Gene
CNPY2
Gene Name
canopy FGF signaling regulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190991.1 477 UTR 5 NP_001177920.1
NM_014255.5 477 UTR 5 NP_055070.1
Gene
PAN2
Gene Name
PAN2 poly(A) specific ribonuclease subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127460.2 477 Intron NP_001120932.1
NM_001166279.1 477 Intron NP_001159751.1
NM_014871.4 477 Intron NP_055686.3

View Full Product Details