Product Details

SNP ID
rs995091576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63600953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGCTGTTGAGCCGCCCCGGGCGG[A/G]CCCAAGCCTTTGGATCTCAGGTGAC
Phenotype
MIM: 605973 MIM: 612958
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCAF7 PubMed Links

Gene Details

Gene
DCAF7
Gene Name
DDB1 and CUL4 associated factor 7
There are no transcripts associated with this gene.

Gene
TACO1
Gene Name
translational activator of cytochrome c oxidase I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016360.3 82 UTR 5 NP_057444.2

View Full Product Details