Product Details

SNP ID
rs55875667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141182528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTAGGATCCTGGGGATACATGAA[A/G]CTTCTGTGAACCAACTTTTCAAGAA
Phenotype
MIM: 606336 MIM: 606345 MIM: 606333 MIM: 606334 MIM: 606335
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDHB10 PubMed Links

Gene Details

Gene
PCDHB10
Gene Name
protocadherin beta 10
There are no transcripts associated with this gene.

Gene
PCDHB16
Gene Name
protocadherin beta 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020957.3 839 UTR 5 NP_066008.2
Gene
PCDHB7
Gene Name
protocadherin beta 7
There are no transcripts associated with this gene.

Gene
PCDHB8
Gene Name
protocadherin beta 8
There are no transcripts associated with this gene.

Gene
PCDHB9
Gene Name
protocadherin beta 9
There are no transcripts associated with this gene.

View Full Product Details