Product Details

SNP ID
rs76286501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125908055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTGTTTATGTTTGGAAGTTGAG[G/T]TATGAATGATCTATCTCCAAATACT
Phenotype
MIM: 607663 MIM: 610693 MIM: 616283
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DDX25 PubMed Links

Gene Details

Gene
DDX25
Gene Name
DEAD-box helicase 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013264.4 Intron NP_037396.3
XM_011542790.2 Intron XP_011541092.1
XM_011542791.2 Intron XP_011541093.1
XM_017017626.1 Intron XP_016873115.1
Gene
HYLS1
Gene Name
HYLS1, centriolar and ciliogenesis associated
There are no transcripts associated with this gene.

Gene
PUS3
Gene Name
pseudouridylate synthase 3
There are no transcripts associated with this gene.

View Full Product Details