Product Details

SNP ID
rs367762890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35150160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACGTGCAGCGTTGTGCCACCCT[C/T]GAGCCACGGAACCAGAACTTCCAGG
Phenotype
MIM: 611220
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NLE1 PubMed Links

Gene Details

Gene
NLE1
Gene Name
notchless homolog 1
There are no transcripts associated with this gene.

Gene
UNC45B
Gene Name
unc-45 myosin chaperone B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033576.1 318 Silent Mutation CTC,CTT L106L NP_001028748.1
NM_001267052.1 318 Silent Mutation CTC,CTT L106L NP_001253981.1
NM_001308281.1 318 Silent Mutation CTC,CTT L106L NP_001295210.1
NM_173167.3 318 Silent Mutation CTC,CTT L106L NP_775259.1
XM_017024234.1 318 Silent Mutation CTC,CTT L106L XP_016879723.1

View Full Product Details