Product Details

SNP ID
rs147411424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154295775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCTGCACCGACATCACATAAAC[G/T]CACTGGGCAGCTCGCAGGCGCCATT
Phenotype
MIM: 300092 MIM: 300044
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TEX28 PubMed Links

Gene Details

Gene
TEX28
Gene Name
testis expressed 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205201.1 102 Intron NP_001192130.1
NM_001586.2 102 Intron NP_001577.1
XM_011531116.1 102 Intron XP_011529418.1
XM_011531117.1 102 Intron XP_011529419.1
XM_011531118.2 102 Intron XP_011529420.1
Gene
TKTL1
Gene Name
transketolase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145933.1 102 UTR 5 NP_001139405.1
NM_001145934.1 102 Intron NP_001139406.1
NM_012253.3 102 UTR 5 NP_036385.3

View Full Product Details