Product Details
- SNP ID
-
rs551479255
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:104747720 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAATGATCCAGGAGCCGCTCCTTT[A/C]CACTCGGGAAACCTTCAGAGGAGTC
- Phenotype
-
MIM: 608871
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NIPSNAP3A
PubMed Links
Gene Details
- Gene
- NIPSNAP3A
- Gene Name
- nipsnap homolog 3A
There are no transcripts associated with this gene.
View Full Product Details