Product Details

SNP ID
rs559244491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3474667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTATCTCTGCATTATAAGCTCT[G/T]AAAGGAAGAACATTCAGTGGGTTTA
Phenotype
MIM: 608034
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ASPA PubMed Links

Gene Details

Gene
ASPA
Gene Name
aspartoacylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000049.2 Intron NP_000040.1
NM_001128085.1 Intron NP_001121557.1
XM_017024661.1 Intron XP_016880150.1
Gene
SPATA22
Gene Name
spermatogenesis associated 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170695.1 Intron NP_001164166.1
NM_001170696.1 Intron NP_001164167.1
NM_001170697.1 Intron NP_001164168.1
NM_001170698.1 Intron NP_001164169.1
NM_001170699.1 Intron NP_001164170.1
NM_001321336.1 Intron NP_001308265.1
NM_001321337.1 Intron NP_001308266.1
NM_032598.4 Intron NP_115987.2

View Full Product Details