Product Details

SNP ID
rs17437516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27188386 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGACGGGTACTCAGCTCCCCAA[C/G]GCTGCGCAATATCCCAGCTTTGCCC
Phenotype
MIM: 142957 MIM: 142958 MIM: 607530 MIM: 142959
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HOXA10 PubMed Links

Gene Details

Gene
HOXA10
Gene Name
homeobox A10
There are no transcripts associated with this gene.

Gene
HOXA10-HOXA9
Gene Name
HOXA10-HOXA9 readthrough
There are no transcripts associated with this gene.

Gene
HOXA11
Gene Name
homeobox A11
There are no transcripts associated with this gene.

Gene
HOXA11-AS
Gene Name
HOXA11 antisense RNA
There are no transcripts associated with this gene.

Gene
HOXA13
Gene Name
homeobox A13
There are no transcripts associated with this gene.

View Full Product Details