Product Details

SNP ID
rs145353484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28080992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGTGAGTCCGGGTTTGGGGATC[C/T]AGATGTCCAGCCCCGTGTCCCCCTC
Phenotype
MIM: 600834
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF165 PubMed Links

Gene Details

Gene
ZNF165
Gene Name
zinc finger protein 165
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003447.3 289 UTR 5 NP_003438.1
XM_017011258.1 289 Intron XP_016866747.1
XM_017011259.1 289 Intron XP_016866748.1
XM_017011260.1 289 Intron XP_016866749.1
XM_017011261.1 289 Intron XP_016866750.1
XM_017011262.1 289 Intron XP_016866751.1
Gene
ZSCAN12P1
Gene Name
zinc finger and SCAN domain containing 12 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details