Product Details

SNP ID
rs59352342
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:57353651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCTGCGATCTTGTTTACTGTTA[C/T]ATTCCTGCTGCGCAGCTCAGGGTCT
Phenotype
MIM: 603343 MIM: 605114
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTRNR2L3 PubMed Links

Gene Details

Gene
MTRNR2L3
Gene Name
MT-RNR2-like 3
There are no transcripts associated with this gene.

Gene
RAE1
Gene Name
ribonucleic acid export 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015885.1 Intron NP_001015885.1
NM_003610.3 Intron NP_003601.1
XM_005260582.2 Intron XP_005260639.2
XM_005260583.2 Intron XP_005260640.1
XM_011529087.2 Intron XP_011527389.1
XM_011529088.2 Intron XP_011527390.1
XM_011529089.1 Intron XP_011527391.1
XM_017028108.1 Intron XP_016883597.1
Gene
SPO11
Gene Name
SPO11, initiator of meiotic double stranded breaks
There are no transcripts associated with this gene.

View Full Product Details