Product Details

SNP ID
hCV27859174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:174748234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATGTGGTCCATCACCATTGCAA[A/C]GTACTTCCACTCTGCCGCCGCCTGG
Phenotype
MIM: 100690
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CHRNA1 PubMed Links

Gene Details

Gene
CHRNA1
Gene Name
cholinergic receptor nicotinic alpha 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000079.3 1342 Missense Mutation GTT,TTT V422F NP_000070.1
NM_001039523.2 1342 Missense Mutation GTT,TTT V447F NP_001034612.1

View Full Product Details