Product Details
- SNP ID
-
rs104894610
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:56594834 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGTCCGTGCACCTCACGGTGCTGC[G/T]GTGGCGCTGTCAGCGGCGCGGGGGC
- Phenotype
-
MIM: 602991
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NOG
PubMed Links
Gene Details
- Gene
- NOG
- Gene Name
- noggin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005450.4 |
1136 |
Missense Mutation |
CGG,CTG |
R204L |
NP_005441.1 |
View Full Product Details