Product Details

SNP ID
rs121912664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7670699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCAGCTCTCGGAACATCTCGAAG[T/C]GCTCACGCCCACGGATCTGCAGCAA
Phenotype
MIM: 191170
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
TP53 PubMed Links

Gene Details

Gene
TP53
Gene Name
tumor protein p53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000546.5 952 Missense Mutation CCC,CTC P337L NP_000537.3
NM_001126112.2 952 Missense Mutation CCC,CTC P337L NP_001119584.1
NM_001126113.2 952 UTR 3 NP_001119585.1
NM_001126114.2 952 UTR 3 NP_001119586.1
NM_001126115.1 952 Missense Mutation CCC,CTC P205L NP_001119587.1
NM_001126116.1 952 UTR 3 NP_001119588.1
NM_001126117.1 952 UTR 3 NP_001119589.1
NM_001126118.1 952 Missense Mutation CCC,CTC P298L NP_001119590.1
NM_001276695.1 952 UTR 3 NP_001263624.1
NM_001276696.1 952 UTR 3 NP_001263625.1
NM_001276697.1 952 Missense Mutation CCC,CTC P178L NP_001263626.1
NM_001276698.1 952 UTR 3 NP_001263627.1
NM_001276699.1 952 UTR 3 NP_001263628.1
NM_001276760.1 952 Missense Mutation CCC,CTC P298L NP_001263689.1
NM_001276761.1 952 Missense Mutation CCC,CTC P298L NP_001263690.1

View Full Product Details