Product Details

SNP ID
rs4490731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:11371240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAATTATTATAACACACTGCAA[A/T]CCATCCTTTTTATAAATGGATAACT
Phenotype
MIM: 612249
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
THSD7A PubMed Links

Gene Details

Gene
THSD7A
Gene Name
thrombospondin type 1 domain containing 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015204.2 9780 UTR 3 NP_056019.1

View Full Product Details